Order Entry
United Kingdom
ContactUsLinkComponent
GSK484 hydrochloride is a selective and reversible peptidylarginine deiminase 4 (PAD4) inhibitor. GSK484 hydrochloride demonstrates high affinity binding to PAD4 with IC50s of 50 nM in the absence of Calcium. In the presence of 2 mM Calcium, notably 1 * 10 mg
Catalog # MDEXHY-100514
Supplier:  MEDCHEMEXPRESS
GSK484 hydrochloride is a selective and reversible peptidylarginine deiminase 4 (PAD4) inhibitor. GSK484 hydrochloride demonstrates high affinity binding to PAD4 with IC50s of 50 nM in the absence of Calcium. In the presence of 2 mM Calcium, notably 1 * 10 mg
Catalog # MDEXHY-100514
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together