Order Entry
United Kingdom
ContactUsLinkComponent
UK-5099 (PF-1005023) is a potent inhibitor of the mitochondrial pyruvate carrier (MPC). UK-5099 (PF-1005023) inhibits pyruvate-depen dent O2 consumption with an IC50 of 50 nM. 1 * 10 mg
Catalog # MDEXHY-15475
Supplier:  MEDCHEMEXPRESS
UK-5099 (PF-1005023) is a potent inhibitor of the mitochondrial pyruvate carrier (MPC). UK-5099 (PF-1005023) inhibits pyruvate-depen dent O2 consumption with an IC50 of 50 nM. 1 * 10 mg
Catalog # MDEXHY-15475
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together