Order Entry
United Kingdom
ContactUsLinkComponent
Rucaparib (AG014699) is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP 1. Rucaparib is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib has the potential for castratio 1 * 50 mg
Catalog # MDEXHY-10617A
Supplier:  MEDCHEMEXPRESS
Rucaparib (AG014699) is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP 1. Rucaparib is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib has the potential for castratio 1 * 50 mg
Catalog # MDEXHY-10617A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together