Order Entry
United Kingdom
ContactUsLinkComponent
GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. GW4869 is an inhibitor of exosome biog enesis/release[1][2][3][4]. 1 * 25 mg
  MDEXHY-19363
GW4869 is a noncompetitive neutral sphingomyelinase (N-SMase) inhibitor with an IC50 of 1 μM. GW4869 is an inhibitor of exosome biog enesis/release[1][2][3][4]. 1 * 25 mg
  MDEXHY-19363
Frequently Bought Together