Order Entry
United Kingdom
ContactUsLinkComponent
Saruparib (AZD5305) is a potent, orally active and selective PARP inhibitor and trapper with IC50 values of 3 nM and 1400 nM for PAR P1 and PARP2, respectively. Saruparib has anti-proliferative activity and inhibits growth in cells with deficiencies 1 * 1 mg
Catalog # MDEXHY-132167
Supplier:  MEDCHEMEXPRESS
Saruparib (AZD5305) is a potent, orally active and selective PARP inhibitor and trapper with IC50 values of 3 nM and 1400 nM for PAR P1 and PARP2, respectively. Saruparib has anti-proliferative activity and inhibits growth in cells with deficiencies 1 * 1 mg
Catalog # MDEXHY-132167
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together