Order Entry
United Kingdom
ContactUsLinkComponent
Cabozantinib is a potent and orally active inhibitor of VEGFR2 and MET, with IC50 values of 0.035, and 1.3 nM, respectively. Cabozan tinib displays strong inhibition of KIT, RET, AXL, TIE2, and FLT3 (IC50=4.6, 5.2, 7, 14.3, and 11.3 nM, respectively) 1 * 10 mg
Catalog # MDEXHY-13016
Supplier:  MEDCHEMEXPRESS
Cabozantinib is a potent and orally active inhibitor of VEGFR2 and MET, with IC50 values of 0.035, and 1.3 nM, respectively. Cabozan tinib displays strong inhibition of KIT, RET, AXL, TIE2, and FLT3 (IC50=4.6, 5.2, 7, 14.3, and 11.3 nM, respectively) 1 * 10 mg
Catalog # MDEXHY-13016
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together