Order Entry
United Kingdom
ContactUsLinkComponent
4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) is an orally active, selective estrogen receptor modulator (SERM). 4-Hydroxytamoxifen (( Z)-4-Hydroxytamoxifen) induces CRISPR/Cas9 systems based on ER mediated nucleus translocation[1][2][3][4]. 1 * 5 mg
Catalog # MDEXHY-16950
Supplier:  MEDCHEMEXPRESS
4-Hydroxytamoxifen ((Z)-4-Hydroxytamoxifen) is an orally active, selective estrogen receptor modulator (SERM). 4-Hydroxytamoxifen (( Z)-4-Hydroxytamoxifen) induces CRISPR/Cas9 systems based on ER mediated nucleus translocation[1][2][3][4]. 1 * 5 mg
Catalog # MDEXHY-16950
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together