Order Entry
United Kingdom
ContactUsLinkComponent
Fedratinib (TG-101348) is a potent, selective, ATP-competitive and orally active JAK2 inhibitor with IC50s of 3 nM for both JAK2 and JAK2V617F kinase. Fedratinib shows 35- and 334-fold selectivity over JAK1 and JAK3, respectively. Fedratinib induces 1 * 100 mg
Catalog # MDEXHY-10409
Supplier:  MEDCHEMEXPRESS
Fedratinib (TG-101348) is a potent, selective, ATP-competitive and orally active JAK2 inhibitor with IC50s of 3 nM for both JAK2 and JAK2V617F kinase. Fedratinib shows 35- and 334-fold selectivity over JAK1 and JAK3, respectively. Fedratinib induces 1 * 100 mg
Catalog # MDEXHY-10409
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together