Order Entry
United Kingdom
ContactUsLinkComponent
Polyinosinic-polycytidylic acid (Poly(I:C)) is a synthetic analog of double-stranded RNA and an agonist of toll-like receptor 3 (TLR 3) and retinoic acid inducible gene I (RIG-I)-like receptors (RIG-I and MDA5). Polyinosinic-polycytidylic acid can be 1 * 10 mg
Catalog # MDEXHY-107202
Supplier:  MEDCHEMEXPRESS
Polyinosinic-polycytidylic acid (Poly(I:C)) is a synthetic analog of double-stranded RNA and an agonist of toll-like receptor 3 (TLR 3) and retinoic acid inducible gene I (RIG-I)-like receptors (RIG-I and MDA5). Polyinosinic-polycytidylic acid can be 1 * 10 mg
Catalog # MDEXHY-107202
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together