Order Entry
United Kingdom
ContactUsLinkComponent
A-485 is a potent and selective catalytic inhibitor of p300/CBP with IC50s of 9.8?nM and 2.6?nM for p300 and CBP histone acetyltrans ferase (HAT), respectively[1]. 1 * 100 mg
Catalog # MDEXHY-107455
Supplier:  MEDCHEMEXPRESS
A-485 is a potent and selective catalytic inhibitor of p300/CBP with IC50s of 9.8?nM and 2.6?nM for p300 and CBP histone acetyltrans ferase (HAT), respectively[1]. 1 * 100 mg
Catalog # MDEXHY-107455
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together