Order Entry
United Kingdom
ContactUsLinkComponent
Dupilumab (REGN-668) is a fully human mAb to IL-4 receptor α (IL-4Rα) that inhibits both IL-4 and IL-13 signaling, markedly improved moderate-to-severe atopic dermatitis[1]. 1 * 1 mg
Catalog # MDEXHY-P9926
Supplier:  MEDCHEMEXPRESS
Dupilumab (REGN-668) is a fully human mAb to IL-4 receptor α (IL-4Rα) that inhibits both IL-4 and IL-13 signaling, markedly improved moderate-to-severe atopic dermatitis[1]. 1 * 1 mg
Catalog # MDEXHY-P9926
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together