Order Entry
United Kingdom
ContactUsLinkComponent
BI-2865 is a none-covalent pan-KRAS Inhibitor. BI-2865 binds to WT, G12C, G12D, G12V and G13D mutant KRAS with KDs of 6.9, 4.5, 32, 26, 4.3 nM respectively. BI-2865 inhibits the proliferation of G12C, G12D or G12V mutant KRAS expressing BaF3 cells (m 1 * 100 mg
Catalog # MDEXHY-153724
Supplier:  MEDCHEMEXPRESS
BI-2865 is a none-covalent pan-KRAS Inhibitor. BI-2865 binds to WT, G12C, G12D, G12V and G13D mutant KRAS with KDs of 6.9, 4.5, 32, 26, 4.3 nM respectively. BI-2865 inhibits the proliferation of G12C, G12D or G12V mutant KRAS expressing BaF3 cells (m 1 * 100 mg
Catalog # MDEXHY-153724
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together