Order Entry
United Kingdom
ContactUsLinkComponent
MHY1485 is a potent cell-permeable mTOR activator that targets the ATP domain of mTOR. MHY1485 inhibits autophagy by suppression of fusion between autophagosomes and lysosomes[1]. 1 * 50 mg
Catalog # MDEXHY-B0795
Supplier:  MEDCHEMEXPRESS
MHY1485 is a potent cell-permeable mTOR activator that targets the ATP domain of mTOR. MHY1485 inhibits autophagy by suppression of fusion between autophagosomes and lysosomes[1]. 1 * 50 mg
Catalog # MDEXHY-B0795
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together