Order Entry
United Kingdom
ContactUsLinkComponent
Ro-3306 is a potent and selective inhibitor of CDK1, with Kis of 20 nM, 35 nM and 340 nM for CDK1, CDK1/cyclin B1 and CDK2/cyclin E, respectively. 1 * 5 mg
  MDEXHY-12529
Ro-3306 is a potent and selective inhibitor of CDK1, with Kis of 20 nM, 35 nM and 340 nM for CDK1, CDK1/cyclin B1 and CDK2/cyclin E, respectively. 1 * 5 mg
  MDEXHY-12529
Frequently Bought Together