Order Entry
United Kingdom
ContactUsLinkComponent
Resiquimod is a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist that induces the upregulation of cytokines such as TNF-α, IL-6 and IF N-α. 1 * 50 mg
Catalog # MDEXHY-13740
Supplier:  MEDCHEMEXPRESS
Resiquimod is a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist that induces the upregulation of cytokines such as TNF-α, IL-6 and IF N-α. 1 * 50 mg
Catalog # MDEXHY-13740
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together