Order Entry
United Kingdom
ContactUsLinkComponent
PolQi2 is a PolΘ inhibitor that targets and inhibits alt-EJ (alternative end-joining) repair by inhibiting the helicase domain at th e N-terminus of PolΘ. PolQi2 enhances the precision and integration efficiency of gene editing at different loci and 1 * 100 mg
  MDEXHY-150279
PolQi2 is a PolΘ inhibitor that targets and inhibits alt-EJ (alternative end-joining) repair by inhibiting the helicase domain at th e N-terminus of PolΘ. PolQi2 enhances the precision and integration efficiency of gene editing at different loci and 1 * 100 mg
  MDEXHY-150279
Frequently Bought Together