Order Entry
United Kingdom
ContactUsLinkComponent
PLX5622 is a highly selective brain penetrant and orally active CSF1R inhibitor (IC50=0.016 μM Ki=5.9 nM). PLX5622 allows for exten ded and specific microglial cells elimination, preceding and during pathology development. PLX5622 demonstrates desir 1 * 100 mg
Catalog # MDEXHY-114153
Supplier:  MEDCHEMEXPRESS
PLX5622 is a highly selective brain penetrant and orally active CSF1R inhibitor (IC50=0.016 μM Ki=5.9 nM). PLX5622 allows for exten ded and specific microglial cells elimination, preceding and during pathology development. PLX5622 demonstrates desir 1 * 100 mg
Catalog # MDEXHY-114153
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together