Order Entry
United Kingdom
ContactUsLinkComponent
Pomalidomide, the third-generation immunomodulatory agent, acts as molecular glue. Pomalidomide interacts with the E3 ligase cereblo n and induces degradation of essential Ikaros transcription factors. 1 * 10 mg
  MDEXHY-10984
Pomalidomide, the third-generation immunomodulatory agent, acts as molecular glue. Pomalidomide interacts with the E3 ligase cereblo n and induces degradation of essential Ikaros transcription factors. 1 * 10 mg
  MDEXHY-10984
Frequently Bought Together