Order Entry
United Kingdom
ContactUsLinkComponent
Revumenib (SNDX-5613) is a potent and specific Menin-MLL inhibitor with a binding Ki of 0.149 nM and a cell based IC50 of 10-20 nM. Revumenib can be used for the research of MLL-rearranged (MLL-r) acute leukemias, including acute lymphoblastic leukem 1 * 5 mg
Catalog # MDEXHY-136175
Supplier:  MEDCHEMEXPRESS
Revumenib (SNDX-5613) is a potent and specific Menin-MLL inhibitor with a binding Ki of 0.149 nM and a cell based IC50 of 10-20 nM. Revumenib can be used for the research of MLL-rearranged (MLL-r) acute leukemias, including acute lymphoblastic leukem 1 * 5 mg
Catalog # MDEXHY-136175
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together