Order Entry
United Kingdom
ContactUsLinkComponent
Sunitinib (SU 11248) is a multi-targeted receptor tyrosine kinase inhibitor with IC50s of 80 nM and 2 nM for VEGFR2 and PDGFRβ, resp ectively[1]. Sunitinib, an ATP-competitive inhibitor, effectively inhibits autophosphorylation of Ire1α by inhibiting 1 * 100 mg
Catalog # MDEXHY-10255A
Supplier:  MEDCHEMEXPRESS
Sunitinib (SU 11248) is a multi-targeted receptor tyrosine kinase inhibitor with IC50s of 80 nM and 2 nM for VEGFR2 and PDGFRβ, resp ectively[1]. Sunitinib, an ATP-competitive inhibitor, effectively inhibits autophosphorylation of Ire1α by inhibiting 1 * 100 mg
Catalog # MDEXHY-10255A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together