Order Entry
United Kingdom
ContactUsLinkComponent
Wortmannin (SL-2052 KY-12420) is a potent, selective and irreversible PI3K inhibitor with an IC50 of 3 nM. Wortmannin also blocks a utophagy formation, and potently inhibits Polo-like kinase 1 (PlK1) and Plk3 with IC50s of 5.8 and 48 nM, respectivel 1 * 100 mg
Catalog # MDEXHY-10197
Supplier:  MEDCHEMEXPRESS
Wortmannin (SL-2052 KY-12420) is a potent, selective and irreversible PI3K inhibitor with an IC50 of 3 nM. Wortmannin also blocks a utophagy formation, and potently inhibits Polo-like kinase 1 (PlK1) and Plk3 with IC50s of 5.8 and 48 nM, respectivel 1 * 100 mg
Catalog # MDEXHY-10197
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together