Order Entry
United Kingdom
ContactUsLinkComponent
RMC-7977 is an orally active triple-complex RAS inhibitor that can simultaneously bind to cyclophilin A (CYPA) (Kd = 195 nM) and KRA S (G12V) (Kd = 292 μM). It exhibits broad-spectrum inhibitory activity against KRAS, NRAS, and HRAS proteins and thei 1 * 10 mg
Catalog # MDEXHY-156498
Supplier:  MEDCHEMEXPRESS
RMC-7977 is an orally active triple-complex RAS inhibitor that can simultaneously bind to cyclophilin A (CYPA) (Kd = 195 nM) and KRA S (G12V) (Kd = 292 μM). It exhibits broad-spectrum inhibitory activity against KRAS, NRAS, and HRAS proteins and thei 1 * 10 mg
Catalog # MDEXHY-156498
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together