Order Entry
United Kingdom
ContactUsLinkComponent
Crizotinib (PF-02341066) is an orally bioavailable, ATP-competitive ALK and c-Met inhibitor with IC50s of 20 and 8 nM, respectively. Crizotinib inhibits tyrosine phosphorylation of NPM-ALK and tyrosine phosphorylation of c-Met with IC50s of 24 and 1 1 * 200 mg
Catalog # MDEXHY-50878
Supplier:  MEDCHEMEXPRESS
Crizotinib (PF-02341066) is an orally bioavailable, ATP-competitive ALK and c-Met inhibitor with IC50s of 20 and 8 nM, respectively. Crizotinib inhibits tyrosine phosphorylation of NPM-ALK and tyrosine phosphorylation of c-Met with IC50s of 24 and 1 1 * 200 mg
Catalog # MDEXHY-50878
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together