Order Entry
United Kingdom
ContactUsLinkComponent
Verteporfin (CL 318952) is a photosensitizer for photodynamic therapy to eliminate the abnormal blood vessels in the eye associated with conditions such as age-related macular degeneration. Verteporfin is a YAP inhibitor which disrupts YAP-TEAD inter 1 * 5 mg
Catalog # MDEXHY-B0146
Supplier:  MEDCHEMEXPRESS
Verteporfin (CL 318952) is a photosensitizer for photodynamic therapy to eliminate the abnormal blood vessels in the eye associated with conditions such as age-related macular degeneration. Verteporfin is a YAP inhibitor which disrupts YAP-TEAD inter 1 * 5 mg
Catalog # MDEXHY-B0146
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together