Order Entry
United Kingdom
ContactUsLinkComponent
Gefitinib (ZD1839) is a potent, selective and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. Gefitinib selectiv ely inhibits EGF-stimulated tumor cell growth (IC50 of 54 nM) and that blocks EGF-stimulated EGFR autophosphorylation 1 * 100 mg
Catalog # MDEXHY-50895
Supplier:  MEDCHEMEXPRESS
Gefitinib (ZD1839) is a potent, selective and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. Gefitinib selectiv ely inhibits EGF-stimulated tumor cell growth (IC50 of 54 nM) and that blocks EGF-stimulated EGFR autophosphorylation 1 * 100 mg
Catalog # MDEXHY-50895
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together