Order Entry
United Kingdom
ContactUsLinkComponent
LY294002 is a broad-spectrum inhibitor of PI3K with IC50s of 0.5, 0.57, and 0.97 μM for PI3Kα, PI3Kδ and PI3Kβ, respectively[1]. LY2 94002 also inhibits CK2 with an IC50 of 98 nM[2]. LY294002 is a competitive DNA-PK inhibitor that binds reversibly to 1 * 500 mg
Catalog # MDEXHY-10108
Supplier:  MEDCHEMEXPRESS
LY294002 is a broad-spectrum inhibitor of PI3K with IC50s of 0.5, 0.57, and 0.97 μM for PI3Kα, PI3Kδ and PI3Kβ, respectively[1]. LY2 94002 also inhibits CK2 with an IC50 of 98 nM[2]. LY294002 is a competitive DNA-PK inhibitor that binds reversibly to 1 * 500 mg
Catalog # MDEXHY-10108
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together