Order Entry
United Kingdom
ContactUsLinkComponent
Imatinib (STI571) is an orally bioavailable tyrosine kinases inhibitor that selectively inhibits BCR/ABL, v-Abl, PDGFR and c-kit kin ase activity. Imatinib (STI571) works by binding close to the ATP binding site, locking it in a closed or self-inhibi 1 * 500 mg
Catalog # MDEXHY-15463
Supplier:  MEDCHEMEXPRESS
Imatinib (STI571) is an orally bioavailable tyrosine kinases inhibitor that selectively inhibits BCR/ABL, v-Abl, PDGFR and c-kit kin ase activity. Imatinib (STI571) works by binding close to the ATP binding site, locking it in a closed or self-inhibi 1 * 500 mg
Catalog # MDEXHY-15463
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together