Order Entry
United Kingdom
ContactUsLinkComponent
Dxd (Exatecan derivative for ADC) is a potent DNA topoisomerase I inhibitor, with an IC50 of 0.31 μM, used as a conjugated drug of H ER2-targeting ADC (DS-8201a). 1 * 1 mg
Catalog # MDEXHY-13631D
Supplier:  MEDCHEMEXPRESS
Dxd (Exatecan derivative for ADC) is a potent DNA topoisomerase I inhibitor, with an IC50 of 0.31 μM, used as a conjugated drug of H ER2-targeting ADC (DS-8201a). 1 * 1 mg
Catalog # MDEXHY-13631D
Supplier:  MEDCHEMEXPRESS