Order Entry
United Kingdom
ContactUsLinkComponent
Upadacitinib (ABT-494) is a potent, orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50=43 nM). Upadacitinib (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific, relev 1 * 100 mg
Catalog # MDEXHY-19569
Supplier:  MEDCHEMEXPRESS
Upadacitinib (ABT-494) is a potent, orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50=43 nM). Upadacitinib (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific, relev 1 * 100 mg
Catalog # MDEXHY-19569
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together