Order Entry
United Kingdom
ContactUsLinkComponent
Nutlin-3a (Rebemadlin), an active enantiomer of Nutlin-3, is a potent murine double minute (MDM2) inhibitor (IC50=90 nM). Nutlin-3a inhibits MDM2-p53?interactions and stabilizes the p53 protein, and induces cell autophagy and apoptosis. Nutlin-3a has 1 * 25 mg
Catalog # MDEXHY-10029
Supplier:  MEDCHEMEXPRESS
Nutlin-3a (Rebemadlin), an active enantiomer of Nutlin-3, is a potent murine double minute (MDM2) inhibitor (IC50=90 nM). Nutlin-3a inhibits MDM2-p53?interactions and stabilizes the p53 protein, and induces cell autophagy and apoptosis. Nutlin-3a has 1 * 25 mg
Catalog # MDEXHY-10029
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together