Order Entry
United Kingdom
ContactUsLinkComponent
FCCP is an uncoupler of oxidative phosphorylation (OXPHOS) in mitochondria. FCCP induces activation of PINK1 leading to Parkin Ser65 phosphorylation[1]. 1 * 50 mg
Catalog # MDEXHY-100410
Supplier:  MEDCHEMEXPRESS
FCCP is an uncoupler of oxidative phosphorylation (OXPHOS) in mitochondria. FCCP induces activation of PINK1 leading to Parkin Ser65 phosphorylation[1]. 1 * 50 mg
Catalog # MDEXHY-100410
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together