Order Entry
United Kingdom
ContactUsLinkComponent
BAY 11-7082 is an IκBα phosphorylation and NF-κB inhibitor. BAY 11-7082 selectively and irreversibly inhibits the TNF-α-induced phos phorylation of IκB-α, and decreases NF-κB and expression of adhesion molecules. BAY 11-7082 inhibits ubiquitin-specif 1 * 100 mg
Catalog # MDEXHY-13453
Supplier:  MEDCHEMEXPRESS
BAY 11-7082 is an IκBα phosphorylation and NF-κB inhibitor. BAY 11-7082 selectively and irreversibly inhibits the TNF-α-induced phos phorylation of IκB-α, and decreases NF-κB and expression of adhesion molecules. BAY 11-7082 inhibits ubiquitin-specif 1 * 100 mg
Catalog # MDEXHY-13453
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together