Order Entry
United Kingdom
ContactUsLinkComponent
Vemurafenib (PLX4032) is a first-in-class, selective, potent inhibitor of B-RAF kinase, with IC50s of 31 and 48 nM for RAFV600E and c-RAF-1, respectively[1][4]. Vemurafenib induces cell autophagy[5]. 1 * 10 mg
Catalog # MDEXHY-12057
Supplier:  MEDCHEMEXPRESS
Vemurafenib (PLX4032) is a first-in-class, selective, potent inhibitor of B-RAF kinase, with IC50s of 31 and 48 nM for RAFV600E and c-RAF-1, respectively[1][4]. Vemurafenib induces cell autophagy[5]. 1 * 10 mg
Catalog # MDEXHY-12057
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together