Order Entry
United Kingdom
ContactUsLinkComponent
Etomoxir ((R)-(+)-Etomoxir) is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidatio n (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig. 1 * 5 mg
Catalog # MDEXHY-50202
Supplier:  MEDCHEMEXPRESS
Etomoxir ((R)-(+)-Etomoxir) is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidatio n (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig. 1 * 5 mg
Catalog # MDEXHY-50202
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together