Order Entry
United Kingdom
ContactUsLinkComponent
RRibociclib (LEE011) is an ATP-competitive and orally active CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex[1][1]. 1 * 100 mg
Catalog # MDEXHY-15777
Supplier:  MEDCHEMEXPRESS
RRibociclib (LEE011) is an ATP-competitive and orally active CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex[1][1]. 1 * 100 mg
Catalog # MDEXHY-15777
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together