Order Entry
United Kingdom
ContactUsLinkComponent
MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active, BBB-penetrated allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy[1][2]. 1 * 5 mg
Catalog # MDEXHY-10358
Supplier:  MEDCHEMEXPRESS
MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active, BBB-penetrated allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy[1][2]. 1 * 5 mg
Catalog # MDEXHY-10358
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together