Order Entry
United Kingdom
ContactUsLinkComponent
Methotrexate (Amethopterin), an antimetabolite and antifolate agent, inhibits the enzyme dihydrofolate reductase, thereby preventing the conversion of folic acid into tetrahydrofolate, and inhibiting DNA synthesis. Methotrexate, also an immunosuppre 1 * 100 mg
Catalog # MDEXHY-14519
Supplier:  MEDCHEMEXPRESS
Methotrexate (Amethopterin), an antimetabolite and antifolate agent, inhibits the enzyme dihydrofolate reductase, thereby preventing the conversion of folic acid into tetrahydrofolate, and inhibiting DNA synthesis. Methotrexate, also an immunosuppre 1 * 100 mg
Catalog # MDEXHY-14519
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together