Order Entry
United Kingdom
ContactUsLinkComponent
Capivasertib (AZD5363) is an orally active and potent pan-AKT kinase inhibitor with IC50 of 3, 7 and 7 nM for Akt1,Akt2 and Akt3, re spectively. 1 * 100 mg
  MDEXHY-15431
Capivasertib (AZD5363) is an orally active and potent pan-AKT kinase inhibitor with IC50 of 3, 7 and 7 nM for Akt1,Akt2 and Akt3, re spectively. 1 * 100 mg
  MDEXHY-15431
Frequently Bought Together