Order Entry
United Kingdom
ContactUsLinkComponent
Everolimus (RAD001) is a Rapamycin (HY-10219) derivative and a potent, selective and orally active mTOR1 inhibitor. Everolimus binds to FKBP-12 to generate an immunosuppressive complex. Everolimus inhibits tumor cells proliferation and induces cell u 1 * 500 mg
Catalog # MDEXHY-10218
Supplier:  MEDCHEMEXPRESS
Everolimus (RAD001) is a Rapamycin (HY-10219) derivative and a potent, selective and orally active mTOR1 inhibitor. Everolimus binds to FKBP-12 to generate an immunosuppressive complex. Everolimus inhibits tumor cells proliferation and induces cell u 1 * 500 mg
Catalog # MDEXHY-10218
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together