Order Entry
United Kingdom
ContactUsLinkComponent
Temozolomide (NSC 362856) is an oral active DNA alkylating agent that crosses the blood-brain barrier. Temozolomide is also a proaut ophagic and proapoptotic agent. Temozolomide is effective against tumor cells that are characterized by low levels of 1 * 100 mg
Catalog # MDEXHY-17364
Supplier:  MEDCHEMEXPRESS
Temozolomide (NSC 362856) is an oral active DNA alkylating agent that crosses the blood-brain barrier. Temozolomide is also a proaut ophagic and proapoptotic agent. Temozolomide is effective against tumor cells that are characterized by low levels of 1 * 100 mg
Catalog # MDEXHY-17364
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together