Order Entry
United Kingdom
ContactUsLinkComponent
Cetuximab (C225) is a human IgG1 monoclonal antibody that inhibits epidermal growth factor receptor (EGFR), with a Kd of 0.201 nM fo r EGFR by SPR. Cetuximab has potent antitumor activity[1]. 1 * 25 mg
Catalog # MDEXHY-P9905
Supplier:  MEDCHEMEXPRESS
Cetuximab (C225) is a human IgG1 monoclonal antibody that inhibits epidermal growth factor receptor (EGFR), with a Kd of 0.201 nM fo r EGFR by SPR. Cetuximab has potent antitumor activity[1]. 1 * 25 mg
Catalog # MDEXHY-P9905
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together