Order Entry
United Kingdom
ContactUsLinkComponent
5-Azacytidine (Azacitidine 5-AzaC Ladakamycin) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation. 5- Azacytidine is incorporated into DNA to covalently trap DNA methyltransferases and contributes to reverse epigenetic 1 * 200 mg
Catalog # MDEXHY-10586
Supplier:  MEDCHEMEXPRESS
5-Azacytidine (Azacitidine 5-AzaC Ladakamycin) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation. 5- Azacytidine is incorporated into DNA to covalently trap DNA methyltransferases and contributes to reverse epigenetic 1 * 200 mg
Catalog # MDEXHY-10586
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together