Order Entry
United Kingdom
ContactUsLinkComponent
Osimertinib (AZD9291) is a covalent, orally active, irreversible, and mutant-selective EGFR inhibitor with an apparent IC50 of 12 nM against L858R and 1 nM against L858R/T790M, respectively. Osimertinib overcomes T790M-mediated resistance to EGFR in 1 * 500 mg
Catalog # MDEXHY-15772
Supplier:  MEDCHEMEXPRESS
Osimertinib (AZD9291) is a covalent, orally active, irreversible, and mutant-selective EGFR inhibitor with an apparent IC50 of 12 nM against L858R and 1 nM against L858R/T790M, respectively. Osimertinib overcomes T790M-mediated resistance to EGFR in 1 * 500 mg
Catalog # MDEXHY-15772
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together