Order Entry
United Kingdom
ContactUsLinkComponent
DAPT (GSI-IX) is a potent and orally active γ-secretase inhibitor with IC50s of 115 nM and 200 nM for total amyloid-β (Aβ) and Aβ42, respectively. DAPT inhibits the activation of Notch 1 signaling and induces cell differentiation. DAPT also induces p 1 * 100 mg
Catalog # MDEXHY-13027
Supplier:  MEDCHEMEXPRESS
DAPT (GSI-IX) is a potent and orally active γ-secretase inhibitor with IC50s of 115 nM and 200 nM for total amyloid-β (Aβ) and Aβ42, respectively. DAPT inhibits the activation of Notch 1 signaling and induces cell differentiation. DAPT also induces p 1 * 100 mg
Catalog # MDEXHY-13027
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together