Order Entry
United Kingdom
ContactUsLinkComponent
Monomethyl auristatin E (MMAE SGD-1010) is a synthetic derivative of dolastatin 10 and functions as a potent mitotic inhibitor by i nhibiting tubulin polymerization. MMAE is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) to 1 * 100 mg
Catalog # MDEXHY-15162
Supplier:  MEDCHEMEXPRESS
Monomethyl auristatin E (MMAE SGD-1010) is a synthetic derivative of dolastatin 10 and functions as a potent mitotic inhibitor by i nhibiting tubulin polymerization. MMAE is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) to 1 * 100 mg
Catalog # MDEXHY-15162
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together