Order Entry
United Kingdom
ContactUsLinkComponent
Dasatinib (BMS-354825) is a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity. The Kis are 16 pM and 30 pM for Src and Bcr-Abl, respectively. Dasatinib inhibits Bcr-Abl and Src with IC50s of [1]. 1 * 100 mg
Catalog # MDEXHY-10181
Supplier:  MEDCHEMEXPRESS
Dasatinib (BMS-354825) is a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity. The Kis are 16 pM and 30 pM for Src and Bcr-Abl, respectively. Dasatinib inhibits Bcr-Abl and Src with IC50s of [1]. 1 * 100 mg
Catalog # MDEXHY-10181
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together