Order Entry
United Kingdom
ContactUsLinkComponent
Z-VAD-FMK (Z-VAD(OH)-FMK) is a well-know pan caspase inhibitor, which does not inhibit ubiquitin carboxy-terminal hydrolase L1 (UCHL 1) activity even at concentrations as high as 440 μM[1]. 1 * 5 mg
Catalog # MDEXHY-16658B
Supplier:  MEDCHEMEXPRESS
Z-VAD-FMK (Z-VAD(OH)-FMK) is a well-know pan caspase inhibitor, which does not inhibit ubiquitin carboxy-terminal hydrolase L1 (UCHL 1) activity even at concentrations as high as 440 μM[1]. 1 * 5 mg
Catalog # MDEXHY-16658B
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together