Order Entry
United Kingdom
ContactUsLinkComponent
Navitoclax (ABT-263) is a potent and orally active Bcl-2 family protein inhibitor that binds to multiple anti-apoptotic Bcl-2 family proteins, such as Bcl-xL, Bcl-2 and Bcl-w, with a Ki of less than 1 nM[1]. 1 * 5 mg
Catalog # MDEXHY-10087
Supplier:  MEDCHEMEXPRESS
Navitoclax (ABT-263) is a potent and orally active Bcl-2 family protein inhibitor that binds to multiple anti-apoptotic Bcl-2 family proteins, such as Bcl-xL, Bcl-2 and Bcl-w, with a Ki of less than 1 nM[1]. 1 * 5 mg
Catalog # MDEXHY-10087
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together