Order Entry
United Kingdom
ContactUsLinkComponent
Palbociclib (PD 0332991) is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbo ciclib has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the 1 * 50 mg
  MDEXHY-50767
Palbociclib (PD 0332991) is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbo ciclib has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the 1 * 50 mg
  MDEXHY-50767
Frequently Bought Together