Order Entry
United Kingdom
ContactUsLinkComponent
Trametinib (GSK1120212 JTP-74057) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametini b activates autophagy and induces apoptosis[1][2]. 1 * 50 mg
Catalog # MDEXHY-10999
Supplier:  MEDCHEMEXPRESS
Trametinib (GSK1120212 JTP-74057) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametini b activates autophagy and induces apoptosis[1][2]. 1 * 50 mg
Catalog # MDEXHY-10999
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together